Learn More
Invitrogen™ Emapalumab Recombinant Monoclonal Antibody
Artikelnummer. 30282448
Change view
Click to view available options
Menge:
1 mg
100 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
Artikelnummer. Menge unitSize
30282448 1 mg 1 Milligramm
30284618 100 μg 100 Mikrogramm
2 options
Artikelnummer. 30282448
Marke: Invitrogen™ MA559498
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Emapalumab, also known as NI-0501, is a fully human monoclonal antibody that targets interferon gamma. Emapalumab development was sponsored by NovImmune SA, further developed by Sobi and FDA approved on November 20, 2018. 1, 5 The approval of emapalumab was followed by the designation of orphan pharmaceutical, priority review and breakthrough therapy. 5 As well, emapalumab was given the status of PRIME by the EMA.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Emapalumab
Anwendungen ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Klassifikation Recombinant Monoclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Gen-Alias emapalumab-lzsg; NI-0501
Wirtsspezies Human
Reinigungsverfahren Protein A
Menge 1 mg
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Lyophilized
Isotype IgG1