Learn More
Invitrogen™ Emapalumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA559497
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Emapalumab, also known as NI-0501, is a fully human monoclonal antibody that targets interferon gamma. Emapalumab development was sponsored by NovImmune SA, further developed by Sobi and FDA approved on November 20, 2018. 1, 5 The approval of emapalumab was followed by the designation of orphan pharmaceutical, priority review and breakthrough therapy. 5 As well, emapalumab was given the status of PRIME by the EMA.
Spezifikation
| Emapalumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| emapalumab-lzsg; NI-0501 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |