missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EML1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP1-30956
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
EML1 Polyclonal specifically detects EML1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
| EML1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| EML1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human EML1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Tested in Human, Mouse, Monkey, and Rat. Predicted cross-reactivity based on sequence identity: Zebrafish (85%). Not yet tested in any other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| echinoderm microtubule associated protein like 1, ELP79, EMAP1, EMAP-1, EMAPechinoderm microtubule-associated protein-like 1, EMAPL1, EMAPLhuEMAP-1, FLJ45033, HuEMAP, HuEMAP-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2009 | |
| Human, Mouse, Rat, Monkey | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur