missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoglin/CD105 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 28117-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Endoglin (Osler-Rendu-Weber syndrome 1), CD105, is a type I membrane glycoprotein located on cell surfaces and is a part of the TGF beta receptor complex. Its gene is mapped to human chromosome 8. The protein consists of a homodimer of 180 kDA with disulfide links. It has been found on endothelial cells, activated macrophages, fibroblasts and smooth muscle cells. Endoglin has a role in the development of the cardiovascular system and in vascular remodeling and has been found to be elevated in pregt women who subsequently develop preeclampsia.Spezifikation
| Endoglin/CD105 | |
| Polyclonal | |
| Unconjugated | |
| ENG | |
| CD105, END, Endoglin, ENG, FLJ41744, HHT1, ORW, ORW1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2022 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P17813 | |
| ENG | |
| Endoglin/CD105 Fusion Protein Ag27822 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur