missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Endothelin 1 Monoclonal Antibody (TR.ET.48.5)
GREENER_CHOICE

Artikelnummer. 11555722
Skift visning
Klik for at se tilgængelige muligheder
Menge:
100 μL
Pakningsstørrelse:
100 Mikroliter
Artikelnummer. Menge unitSize
11555722 100 μL 100 Mikroliter
1 options
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 11555722

Brand: Invitrogen™ MA3005

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

MA3-005 detects ET-1 from human, sheep, mouse and rat tissues. This antibody shows little cross-reactivity to related peptides and no known cross-reactivity to non-related peptides. MA3-005 has been successfully used in Western blot, flow cytometry, immunofluorescence, immunohistochemistry, immunocytochemistry, and radioimmune assay procedures. Immunohistochemical staining of ET-1 in human corpus cavernosum tissue with MA3-005 results in staining of endothelial cells. Radioimmune assays using MA3-005 can be used to concentrate ET-1 in solution (e.g. serμm/plasma, milk, urine). This antibody is not recommended for human placenta tissue or HepG2 cells in Western blot applications. For immunohistochemistry with MA3-005 formalin-fixed, paraffin-embedded tissue sections has been used. The MA3-005 antigen is purified human endothelin-1. Studies suggest that this antibody binds to an epitope in the region of ET-1 represented by amino acids 8-16.

The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Endothelin 1
Anwendungen Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot, Immunocytochemistry
Klassifikation Monoclonal
Klon TR.ET.48.5
Konzentration 2.5 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.05% sodium azide
Gen EDN1
Gen-Zugriffsnummer P05305, P22387, P22388, Q9BG76
Gen-Alias ARCND3; Big endothelin 1; Big endothelin-1; Big ET; Big ET 1; Big ET1; BigET; EDN1; endothelin 1; endothelin-1; Et; ET1; ET-1; HDLCQ7; PPET1; ppET-1; preproendothelin; preproendothelin-1; preproET; QME
Gensymbole EDN1
Wirtsspezies Mouse
Immunogen Pure endothelin-1 conjugated to KLH.
Reinigungsverfahren Affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 13614, 1906, 24323, 443498
Zielspezies Human, Mouse, Sheep, Rat
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG1
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.