Learn More
Invitrogen™ Endothelin B Receptor Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA3066
Beschreibung
PA3-066 detects Endothelin Receptor B from human, mouse, and rat samples. PA3-066 has been successfully used in Western blot, immunocytochemistry, and immunohistochemistry (paraffin) procedures. By Western blot, this antibody detects a 47 kDa protein corresponding to mouse Endothelin Receptor B. The PA3-066 antigen is the COOH-terminal amino acids of mouse ETB (424-442) KANDHGYDNFRSSNKYSSS coupled to KLH.
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
Spezifikation
| Endothelin B Receptor | |
| Polyclonal | |
| Unconjugated | |
| EDNRB | |
| ABCDS; AU022549; Ednra; Ednrb; endothelin B receptor; endothelin receptor non-selective type; endothelin receptor subtype B1; endothelin receptor type B; endothelin-B receptor; ETb; ET-B; Etb receptor; Et-b receptor; ETB1; ETBR; ET-BR; Et-b-r; Etb-svr; ETRB; ET-RB; ETR-b; HSCR; HSCR2; piebald; RP11-318G21.1; Sox10m1; WS4A | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 0.05% sodium azide | |
| P21451, P24530, P48302 | |
| EDNRB | |
| COOH-terminal amino acids of mouse ETB (424-442) KANDHGYDNFRSSNKYSSS coupled to KLH. | |
| RUO | |
| 13618, 1910, 50672 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.