Learn More
Invitrogen™ Endothelin B Receptor Recombinant Rabbit Monoclonal Antibody (24GB3375)
Beschreibung
Endothelin B Receptor Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
Spezifikation
Spezifikation
| Antigen | Endothelin B Receptor |
| Anwendungen | Flow Cytometry, Western Blot, Immunocytochemistry |
| Klassifikation | Recombinant Monoclonal |
| Klon | 24GB3375 |
| Konzentration | 0.8 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | EDNRB |
| Gen-Zugriffsnummer | P21451, P24530, P48302 |
| Gen-Alias | ABCDS; AU022549; Ednra; Ednrb; endothelin B receptor; endothelin receptor non-selective type; endothelin receptor subtype B1; endothelin receptor type B; endothelin-B receptor; ETb; ET-B; Etb receptor; Et-b receptor; ETB1; ETBR; ET-BR; Et-b-r; Etb-svr; ETRB; ET-RB; ETR-b; HSCR; HSCR2; piebald; RP11-318G21.1; Sox10m1; WS4A |
| Mehr anzeigen |