Beschreibung
Enteropeptidase/Enterokinase Polyclonal antibody specifically detects Enteropeptidase/Enterokinase in Bovine samples. It is validated for ELISA
Spezifikation
Spezifikation
| Antigen | Enteropeptidase/Enterokinase |
| Anwendungen | ELISA |
| Klassifikation | Polyclonal |
| Immunogen | Produced in rabbits immunized with purified, recombinant Bovine Enteropeptidase/Enterokinase (Accession#: NP_776864.1; Ile801-His1035). |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Menge | 0.1 mL |
| Zielspezies | Bovine |