missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EOMES Monoclonal Antibody (Dan11mag), PE-Cyanine5, eBioscience™, Invitrogen™
Beschreibung
Rat Monoclonal Antibody
TBR2 is a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar protein disrupted in mice is shown to be essential during trophoblast development and gastrulation. TBR2 plays a role in brain development and is required for the specification and the proliferation of the intermediate progenitor cells and their progeny in the cerebral cortex.
Spezifikation
Spezifikation
| Antigen | EOMES |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | Dan11mag |
| Konzentration | 0.2 mg/mL |
| Konjugat | PE-Cyanine5 |
| Zusammensetzung | PBS with 0.09% sodium azide; pH 7.2 |
| Gen | Eomes |
| Gen-Zugriffsnummer | O54839 |
| Gen-Alias | C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2 |
| Mehr anzeigen |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?