Learn More
Invitrogen™ Epcoritamab Chimeric Recombinant Human Monoclonal Antibody
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Epcoritamab (DuoBody™-CD3xCD20) is a proprietary bispecific antibody created using Genmab's DuoBody technology. It targets CD20 on the B-cells, a clinically well-validated target that is expressed in a wide variety of B-cell malignancies. Epcoritamab is currently under investigation in a Phase I/II study for multiple hematological B cell malignancies.
Spezifikation
Spezifikation
| Antigen | Epcoritamab Chimeric |
| Anwendungen | ELISA, Flow Cytometry, Functional Assay |
| Klassifikation | Recombinant Monoclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Gen-Alias | GEN3013 |
| Wirtsspezies | Human |
| Reinigungsverfahren | Protein A |
| Menge | 100 μg |
| Mehr anzeigen |