missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EphB2 Monoclonal Antibody (512001)

Rat Monoclonal Antibody
Marke: Invitrogen™ MA561467
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
EPHB2 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.
Spezifikation
| EphB2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P54763 | |
| EPHB2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphB2, Val27-Lys548 (predicted). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 512001 | |
| Unconjugated | |
| EPHB2 | |
| CAPB; cEK5; chicken embryo kinase 5; chicken embryo kinase 5 protein; developmentally-regulated Eph-related tyrosine kinase; DRT; EFL6; EFNB3; EK5; ELK; ELK-related tyrosine kinase; embryo kinase 5 protein CEK5; EPH B2; EPH receptor B2; eph tyrosine kinase 3; EPHB2; EphB2/CTF1; EphB2/CTF2; EPHB3; EPH-like kinase 5; Ephrin B1; Ephrin B2; Ephrin B3; Ephrin B4; Ephrin type-B receptor 2; EphrinB1; EphrinB2; EphrinB3; EphrinB4; EPHT3; EPLG8; EPTH3; ERK; ETECK; ETK2; HEK2; Hek5; HEK-6; LERK8; MGC87492; NET; Neural kinase; Nuk; Nuk receptor tyrosine kinase; PCBC; Prkm5; protein-tyrosine kinase HEK5; Qek5; Renal carcinoma antigen NY-REN-47; RGD1564232; Sek3; TYRO5; TYRO6; Tyrosine-protein kinase receptor EPH-3; tyrosine-protein kinase receptor SEK-3; Tyrosine-protein kinase TYRO5 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 13844 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur