Learn More
Invitrogen™ EphB6 Monoclonal Antibody (92224)
GREENER_CHOICE
Artikelnummer. 30534333
Change view
Click to view available options
Menge:
500 μg
Packungsgröße:
500 Mikrogramm
Artikelnummer. Menge unitSize
30534333 500 μg 500 Mikrogramm
1 options
Artikelnummer. 30534333
Marke: Invitrogen™ MA560981
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rat Monoclonal Antibody
Reconstitute at 0.5 mg/mL in sterile PBS.
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The ephrin receptor encoded by this gene lacks the kinase activity of most receptor tyrosine kinases and binds to ephrin-B ligands.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen EphB6
Anwendungen Western Blot
Klassifikation Monoclonal
Klon 92224
Konzentration 0.5 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with trehalose and no preservative
Gen EPHB6
Gen-Zugriffsnummer O08644
Gen-Alias Cekl; chicken eph/elk receptor-like protein; EPB6; EPH receptor B6; EPHB6; EphB6v; Epherin B6; Epherin receptor B6; ephrin type A receptor; ephrin type-B receptor 6; HEP; human kinase-defective Eph-family receptor protein; MEP; MGC129910; MGC129911; Tyrosine-protein kinase-defective receptor EPH-6
Gensymbole EPHB6
Wirtsspezies Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphB6, Leu33-Ser587.
Reinigungsverfahren Protein A/G
Menge 500 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 13848
Zielspezies Mouse
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Lyophilized
Isotype IgG2a