missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Eptinezumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA559139
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Eptinezumab is a fully-humanized IgG1 antibody manufactured using yeast (pichia pastoris) and developed by Lundbeck Seattle Biopharmaceuticals. 7 Eptinezumab has been specifically designed to bind to both alpha and beta forms of the human calcitonin gene-related peptide (CGRP). 8, 1, 2, 3 It was approved by the FDA in February 2020 for the preventive treatment of migraine headaches in adults.
Spezifikation
| Eptinezumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ALD-403 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur