Learn More
Invitrogen™ Estrogen Receptor alpha (ER alpha) ER Alpha Recombinant Protein
Beschreibung
Beinhaltet
One tube of full-length ER Alpha Recombinant Protein.
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. ER (Estrogen Receptor) Alpha Recombinant Protein is an insect cell-expressed, full-length, untagged, human nuclear hormone receptor that binds [3H]-estradiol with sub-nM affinity in hydroxylapatite assays and binds specifically to estrogen response element (ERE) sequences. ER alpha full-length recombinant protein is: Flexible-use in your in-house assay or as part of our PolarScreen™ ER Alpha Competitor assays for a complete assay solution Ready to use-active, purified, and native (untagged) Convenient-available in bulk sizes ER Alpha Recombinant Protein is manufactured under the strictest quality parameters. This product is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond. Used in:PolarScreen™ ER Alpha Competitor Assay, RedPolarScreen™ ER Alpha Competitor Assay, Green Protein Form: Recombinant, Full Length.
Spezifikation
Spezifikation
| Zugriffsnummer | P03372 |
| Konzentration | See Label |
| Zur Verwendung mit (Anwendung) | Kinase Assay |
| Zusammensetzung | Proprietary buffer with no preservative |
| Gen-ID (Entrez) | 2099 |
| Molekulargewicht | 66.4 kDa |
| Name | Estrogen Receptor alpha (ER alpha) ER Alpha |
| Reinigungsverfahren | Purified |
| Menge | 140 μg |
| Lagerungsbedingungen | -80°C, Avoid Freeze/Thaw Cycles |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.