Learn More
Tocris Bioscience™ ERK5-IN-1
Beschreibung
ERK5-IN-1 is a potent and selective ERK5 inhibitor; inhibits EGFR-induced ERK5 autophosphorylation (EC50 = 90 nM) and ERK5 enzymatic activity (IC50 = 162 nM). Exhibits at least 30-fold selectivity for ERK5 over LRRK2 in a cell based assay. Also selective for ERK5 over a panel of other kinases. Orally bioavailable.
Spezifikation
Spezifikation
| Reinheit | >98% |
| Beschreibung | ERK5-IN-1 |
| Molekulargewicht | 638.8 |
| Produkttyp | ERK5-IN-1 |
| Menge | 10 mg |
Nur für Forschungszwecke