missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
ERp72 is an endoplasmic reticulum luminal protein that is both a stress protein and a member of the protein disulfide isomerase family of proteins. Sequence analysis of ERp72 shows that it shares sequence identity with PDI (protein disulphide isomerase) at three discrete regions, as well as containing the ER retention signal KEEL at its C-terminus. ERp72 is shown to have three copies of sequence believed to be CGHC-containing active sites, which are also found in PDI. These CGHC sites are critical sites that are involved in the protein disulphide isomerization process. These results suggest that ERp72 is a newly described member of the family of CGHC-containing proteins that functions as a molecular chaperone.
Spezifikation
Spezifikation
| Antigen | ERp72 |
| Anwendungen | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
| Klassifikation | Monoclonal |
| Klon | 1D5F3 |
| Konzentration | 0.48 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | PDIA4 |
| Gen-Zugriffsnummer | P13667 |
| Gen-Alias | ER protein 70, ER protein 72, ERp 72, ERP70, ERp72, PDIA4, Protein disulfide isomerase A4 |
| Mehr anzeigen |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?