missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin-d6, TRC

Product Code. 30374031
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
1 mg
10 mg
missing translation for 'unitSize'
1 Milligramm
10 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30374031 1 mg 1 Milligramm
30369385 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30374031 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCE6499531MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Erythromycin-d6
CAS 959119-25-6
Summenformel C37H61D6NO13
Menge 1 mg
Synonym E-Base-d6, E-Mycin-d6, Erytromycin A-d6, Aknemycin-d6, Aknin-d6, Eemgel-d6, Ery-Derm-d6, Erymax-d6, Ery-Tab-d6, N,N-didemethyl-N,N-di(methyl-d3)erythromycin
InChI-Formel InChI=1S/C37H67NO13/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1/i11D3,12D3
SMILES [2H]C([2H])([2H])N([C@H]1C[C@@H](C)O[C@@H](O[C@@H]2[C@@H](C)[C@H](O[C@H]3C[C@@](C)(OC)[C@@H](O)[C@H](C)O3)[C@@H](C)C(=O)O[C@H](CC)[C@@](C)(O)[C@H](O)[C@@H](C)C(=O)[C@H](C)C[C@@]2(C)O)[C@@H]1O)C([2H])([2H])[2H]
Molekulargewicht (g/mol) 739.96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.