missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-3-(2-methyl-[1,3]dioxolan-2-yl)propanoat, TRC

Product Code. 30382159
Change view
Click to view available options
Menge:
2.5 g
Unit Size:
2.50 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30382159 2.5 g 2.50 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382159 Supplier Toronto Research Chemicals Supplier No. TRCE9250862.5G
 Eingeschränkter Artikel
Verfügbar ab: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Ethyl 3-(2-methyl-[1,3]dioxolan-2-yl)propanoate
CAS 941-43-5
Summenformel C9H16O4
Menge 2.5 g
Synonym 2-Methyl-1,3-dioxolane-2-propanoic Acid Ethyl Ester, Ethyl levulinate ethylene ketal, Ethyl 4-Dioxolan Pentanoate, NSC 46288
InChI-Formel InChI=1S/C9H16O4/c1-3-11-8(10)4-5-9(2)12-6-7-13-9/h3-7H2,1-2H3
SMILES CCOC(=O)CCC1(C)OCCO1
IUPAC-Name Ethyl-3-(2-methyl-1,3-dioxolan-2-yl)propanoat
Molekulargewicht (g/mol) 188.22
Formelmasse 188.1049
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.