missing translation for 'onlineSavingsMsg'
Learn More

Ethyl-5-nitro-2-furoat, 97 %, Thermo Scientific Chemicals

Artikelnummer. 11446656
Ansicht ändern
Click to view available options
Menge:
1 g
5 g
25 g
Unit Size:
1 Gramm
25 Gramm
5 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
11446656 5 g 5 Gramm
11436656 1 g 1 Gramm
11456656 25 g 25 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446656 Supplier Thermo Scientific Alfa Aesar Supplier No. B24861.06
 Eingeschränkter Artikel
Verfügbar ab: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 943-37-3 | C7H7NO5 | 185.135 g/mol

Dieses Markenprodukt von Thermo Scientific Chemicals war ursprünglich Teil des Produktportfolios von Alfa Aesar. Einige Dokumente und Etiketteninformationen enthalten möglicherweise Angaben zur bisherigen Marke. Die ursprüngliche Produkt- oder Artikelnummer bei Alfa Aesar oder die SKU-Referenz wurde im Rahmen des Übergangs der Marke zu Thermo Scientific Chemicals nicht geändert.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 943-37-3
Summenformel C7H7NO5
Molekulargewicht (g/mol) 185.135
MDL-Nummer MFCD00092566
InChI-Schlüssel JMNXLAQKIHVFIC-UHFFFAOYSA-N
PubChem CID 70341
IUPAC-Name Ethyl 5-Nitrofuran-2-Carboxylat
SMILES CCOC(=O)C1=CC=C(O1)[N+](=O)[O-]

Specifications

Chemischer Name oder Material Ethyl 5-nitro-2-furoate
Schmelzpunkt 98°C to 101°C
Menge 5 g
Synonym ethyl 5-nitro-2-furoate, 2-furoic acid, 5-nitro-, ethyl ester, 5-nitro-furan-2-carboxylic acid ethyl ester, 2-furancarboxylic acid, 5-nitro-, ethyl ester, ethyl5-nitro-2-furoate, acmc-20aky3, ethyl-5-nitro-2-furoate, ethyl 5-nitro-2-furancarboxylate, 5-nitro-2-furoic acid ethyl ester, 5-nitro-2-furancarboxylic acid ethyl ester
Formelmasse 185.14
Reinheit (%) 97%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.