Learn More
Novus Biologicals™ Recombinant Rat SCF/c-kit Ligand Protein
Artikelnummer. 18602506 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
1 mg
10 μg
100 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
500 Mikrogramm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18602506 10 μg 10 Mikrogramm
18684466 100 μg 100 Mikrogramm
18625905 500 μg 500 Mikrogramm
18698215 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 18602506 Lieferant Novus Biologicals™ Lieferanten-Nr. NBP23528810ug
Please to purchase this item. Need a web account? Register with us today!
Recombinant Protein
SCF Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine MC/9-2 mast cells is less than 5 ng/mL, corresponding to a specific activity of > 2.0 x 10^5 IU/mg.
TRUSTED_SUSTAINABILITY
Spezifikation
Zugriffsnummer P21581, P21581, P21581
Zur Verwendung mit (Anwendung) SDS-PAGE, Bioactivity
Zusammensetzung Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4.
Gen-ID (Entrez) 4254
Molekulargewicht MolecularWeight-theroretical: 18.5 kDa
Menge 10 μg
Lagerungsbedingungen Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin-Konzentration Less than 1 EU/μg of SCF/c-kit Ligand as determined by LAL method.
Gen-Alias DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor
Gensymbol KITLG
Rekonstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Forschungskategorie Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Reinheits- oder Qualitätsgrad >95%, by SDS-PAGE and HPLC
Protein SCF/c-kit Ligand