missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Alfa Aesar™ Ethyl hydrogen fumarate, 97%
Beskrivning
Applications
Ethyl hydrogen fumarate was used in the preparation of photo-crosslinkable macromers. It was also used to synthesize Ugi/intramolecular Diels-Alder (IMDA) cycloaddition products.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Ethyl hydrogen fumarate was used in the preparation of photo-crosslinkable macromers. It was also used to synthesize Ugi/intramolecular Diels-Alder (IMDA) cycloaddition products.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifikationer
Specifikationer
| Chemischer Name oder Material | Ethyl hydrogen fumarate |
| CAS | 2459-05-4 |
| Summenformel | C6H8O4 |
| MDL-Nummer | MFCD00002699 |
| Menge | 500g |
| Synonym | monoethyl fumarate, e-4-ethoxy-4-oxobut-2-enoic acid, ethyl hydrogen fumarate, fumaric acid monoethyl ester, e-4-ethoxy-4-oxo-2-butenoic acid, unii-y3w849ng2n, 2e-4-ethoxy-4-oxobut-2-enoic acid, fumaric acid, monoethyl ester, mono-ethyl fumarate, ethyl hydrogen maleate |
| InChI-Schlüssel | XLYMOEINVGRTEX-ONEGZZNKSA-N |
| SMILES | CCOC(=O)C=CC(=O)O |
| IUPAC-Name | (E)-4-ethoxy-4-oxobut-2-enoic acid |
| Molekulargewicht (g/mol) | 144.13 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?