missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Etrolizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559306
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Etrolizumab has been used in trials studying the treatment of Crohn Disease and Ulcerative Colitis.
Tekniske data
| Etrolizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRO145223; RG7413; rhuMAb Beta7 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion