Learn More
Gibco™ Expi293™ MembranePro™ Expression System
Expi293™ MembranePro™ Expression System
N.V.
Spezifikation
Typ | Expressionssystem |
---|---|
Liefertyp | Virusartige Partikel (VLPs) |
Zelllinie | Expi293F™ |
Zur Verwendung mit (Anwendung) | Membranproteinexpression |
Promoter | EF-1a |
Beschreibung
For researchers studying G protein–coupled receptors (GPCRs) and other membrane proteins, the Expi293™ MembranePro™ Expression System delivers enriched, functional membrane proteins with high efficiency, scalability and reliability for a wide variety of low or high throughput applications.Features of the Expi293™ MembranePro™ Expression System include:
In addition, benefits of MembranePro™ technology include:
With the less labor-intensive Expi293™ MembranePro™ Expression System protocol, you will:
1. Clone your gene of interest using a TOPO™ vector
2. Transfect a susupension culture of Expi293F™ cells using Expifectamine™ 293 transfection reagent
3. Harvest lipoparticles 48 hrs after addition of enhancers using MembranePro™ Precipitation Mix
4. Store lipoparticles at -80 C or proceed to downstream assays
Note: Exi293F™ Cells (A14527) and a pEF6 V5-His TOPO™ Expression Kit (K9610-20) are required for use of this system and must be purchased separately when needed.
Spezifikation
Expressionssystem | |
Expi293F™ | |
EF-1a |
Virusartige Partikel (VLPs) | |
Membranproteinexpression |
For Research Use Only. Not for use in diagnostic procedures.