missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, Qdot™ 565
F(ab')2-Goat Polyclonal Secondary Antibody
343.00 € - 484.00 €
Spezifikation
| Antigen | Mouse IgG (H+L) |
|---|---|
| Inhalt und Lagerung | 4°C, store in dark |
| Anwendungen | Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konjugat | Qdot 565 |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
10262783
|
Invitrogen™
Q11031MP |
200 μL |
484.00 €
200 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
10170533
|
Invitrogen™
Q11032MP |
100 μL |
343.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
| Mouse IgG (H+L) | |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Qdot 565 | |
| Goat | |
| RUO | |
| 0.05M borate with 1M betaine and 0.05% sodium azide; pH 8.3 | |
| Secondary | |
| Purified |
| 4°C, store in dark | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Gamma Immunoglobins Heavy and Light chains. | |
| Secondary Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts