missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, FITC
F(ab')2-Goat Polyclonal Secondary Antibody
Marke: Invitrogen™ A24532
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Rabbit IgG (H+L) | |
| Polyclonal | |
| FITC | |
| Goat | |
| Affinity Chromatography | |
| RUO | |
| Rabbit | |
| Secondary Antibody | |
| IgG |
| Immunocytochemistry | |
| 1.5 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.8 | |
| Gamma Immunoglobins Heavy and Light chains. | |
| 500 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur