missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ F4/80 Monoclonal Antibody (Cl:A3-1)

Artikelnummer. p-7172908
Change view
Click to view available options
Menge:
100 μg
250 μg
500 μg
Packungsgröße:
100 Mikrogramm
250 Mikrogramm
500 Mikrogramm
Artikelnummer. Menge unitSize
15245899 500 μg 500 Mikrogramm
15255899 100 μg 100 Mikrogramm
15275899 250 μg 250 Mikrogramm
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 15245899

Marke: Invitrogen™ MA516629

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rat Monoclonal Antibody

This product requires pre-treatment of paraffin sections prior to staining. Proteinase K is recommended for tissues fixed for less than 24 hrs. Citrate buffer (pH 6.0) is recommended for tissues fixed for more than 24 hrs. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti mouse F4/80 antibody, clone Cl:A3-1 recognizes the murine F4/80 antigen, a approximately 160 kDa cell surface glycoprotein member of the EGF-TM7 family of proteins which shares 68% overall amino acid identity with human EGF module-containing mucin-like hormone receptor 1 (EMR1). Rat anti mouse F4/80 antibody, clone Cl:A3-1 recognizes the murine F4/80 antigen, a approximately 160 kDa cell surface glycoprotein member of the EGF-TM7 family of proteins which shares 68% overall amino acid identity with human EGF module-containing mucin-like hormone receptor 1 (EMR1). Rat anti mouse F4/80 antibody, clone Cl:A3-1 recognizes the murine F4/80 antigen, a approximately 160 kDa cell surface glycoprotein member of the EGF-TM7 family of proteins which shares 68% overall amino acid identity with human EGF module-containing mucin-like hormone receptor 1 (EMR1).

F4/80 antigen is a 160 kDa cell surface glycoprotein that is a member of the EGF TM7 family of proteins and shares 68% overall amino acid identity with human EGF module containing mucin like hormone receptor 1 (EMR1). Expression of F4/80 is heterogeneous and is reported to vary during macrophage maturation and activation. The F4/80 antigen is expressed on a wide range of mature tissue macrophages including Kupffer cells, Langerhans, microglia, macrophages located in the gut lamina propria, peritoneal cavity, lung, thymus, bone marrow stroma and macrophages in the red pulp of the spleen. F4/80 expression has also been reported on a subpopulation of dendritic cells but is absent from macrophages located in T cell areas of the spleen and lymph node. The ligands and biological functions of the F4/80 antigen have not yet been determined but recent studies suggest a role for F4/80 in the generation of efferent CD8+ve regulatory T cells.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen F4/80
Anwendungen Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry
Klassifikation Monoclonal
Klon Cl:A3-1
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.09% sodium azide
Gen ADGRE1
Gen-Zugriffsnummer Q61549
Gen-Alias ADGRE1; Adhesion G protein-coupled receptor E1; Adhesion G protein-coupled receptor E2; cell surface glycoprotein EMR1; cell surface glycoprotein F4/80; DD7A5-7; EGF-like module containing, mucin-like, hormone receptor-like 1; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-TM7; Emr1; EMR1 hormone receptor; F4/80; Gpf480; Ly71; lymphocyte antigen 71; TM7LN3
Gensymbole ADGRE1
Wirtsspezies Rat
Immunogen Thioglycollate stimulated peritoneal macrophages from C57/BL mice.
Reinigungsverfahren Protein G
Menge 500 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 13733
Zielspezies Mouse
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG2b
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.