missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XIIIa Mouse anti-Human, Mouse, Porcine, Clone: 2D2B2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66325-1-IG-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene encodes the coagulation factor XIII A subunit. Coagulation factor XIII is the last zymogen to become activated in the blood coagulation cascade. Plasma factor XIII is a heterotetramer composed of 2 A subunits and 2 B subunits. The A subunits have catalytic function, and the B subunits do not have enzymatic activity and may serve as plasma carrier molecules. Platelet factor XIII is comprised only of 2 A subunits, which are identical to those of plasma origin. Upon cleavage of the activation peptide by thrombin and in the presence of calcium ion, the plasma factor XIII dissociates its B subunits and yields the same active enzyme, factor XIIIa, as platelet factor XIII. This enzyme acts as a transglutaminase to catalyze the formation of gamma-glutamyl-epsilon-lysine crosslinking between fibrin molecules, thus stabilizing the fibrin clot. It also crosslinks alpha-2-plasmin inhibitor, or fibronectin, to the alpha chains of fibrin. Factor XIII deficiency is classified into two categories: type I deficiency, characterized by the lack of both the A and B subunits; and type II deficiency, characterized by the lack of the A subunit alone. These defects can result in a lifelong bleeding tendency, defective wound healing, and habitual abortion.Spezifikation
| Factor XIIIa | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00488, Q8BH61 | |
| F13a1 | |
| Factor XIIIa Fusion Protein Ag11105 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Porcine | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2D2B2 | |
| Unconjugated | |
| F13A1 | |
| Coagulation factor XIIIa, F13A, F13A1, Factor XIIIA, Transglutaminase A chain | |
| Mouse | |
| Protein A | |
| RUO | |
| 100153504, 2162, 74145 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur