missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: PSIHSLLYQL NQQCQAPGAA PPACQGMAIP HPSPAKHGPV PSFPSMAYSA AAGLPDCRKG TELGQGATQA LTLAGAAKPA GYADSGLDYL LWPQKP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
Specifications
Specifications
| Antigen | FAM222A |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konzentration | 0.2 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gen | FAM222A |
| Gen-Zugriffsnummer | Q5U5X8 |
| Gen-Alias | C12orf34; FAM222A; family with sequence similarity 222 member A; family with sequence similarity 222, member A; protein FAM222A |
| Gensymbole | FAM222A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
