Learn More
Thermo Scientific™ FastDigest™ Value Pack PROMO
FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in a single buffer.
Marke:  Thermo Scientific™ K1991-RMD123
Artikelnummer. 15619484
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
The Thermo Scientific FastDigest Value Pack contains the 13 most commonly used FastDigest enzymes. The selection of enzymes will support a wide range of molecular biology applications such as vector and insert preparation for cloning, fast clone analysis and digestion of PCR products. Each enzyme in the pack allows for 20 standard restriction digest reactions.
Thermo Scientific FastDigest enzymes are an advanced line of restriction enzymes that are all 100% active in a single buffer. The universal FastDigest and FastDigest Green buffer allows single-, double- or multiple DNA digestion within 5-15 minutes, eliminating any need for buffer change or subsequent DNA clean-up steps. Thermo Scientific DNA modifying enzymes have 100% activity in this buffer as well. For additional convenience the FastDigest Green Buffer includes a density reagent and two tracking dyes which allows for direct loading of digestion reaction products on gels.
The Thermo Scientific FastDigest Value Pack contains the 13 most commonly used FastDigest enzymes. The selection of enzymes will support a wide range of molecular biology applications such as vector and insert preparation for cloning, fast clone analysis and digestion of PCR products. Each enzyme in the pack allows for 20 standard restriction digest reactions.
Thermo Scientific FastDigest enzymes are an advanced line of restriction enzymes that are all 100% active in a single buffer. The universal FastDigest and FastDigest Green buffer allows single-, double- or multiple DNA digestion within 5-15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. Thermo Scientific DNA modifying enzymes have 100% activity in this buffer as well. For additional convenience the FastDigest Green Buffer includes a density reagent and two tracking dyes which allows for direct loading of digestion reaction products on gels.
Highlights
• 100% activity of all FastDigest enzymes in a single buffer
• Complete digestion in 5-15 minutes and direct loading on gels
• 100% buffer compatibility with downstream applications
• 176 FastDigest enzymes available
• No star activity
Includes
FastDigest BglII 20 rxns
FastDigest BamHI 20 rxns
FastDigest EcoRI 20 rxns
FastDigest Eco32I 20 rxns
FastDigest HindIII 20 rxns
FastDigest KpnI 20 rxns
FastDigest NcoI 20 rxns
FastDigest NdeI 20 rxns
FastDigest NotI 20 rxns
FastDigest PstI 20 rxns
FastDigest SmaI 20 rxns
FastDigest XbaI 20 rxns
FastDigest XhoI 20 rxns
10X FastDigest Buffer 2x 1 mL
10X FastDigest Green Buffer 2x 1 mL
Note
The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer. For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.
Definition of activity unit:1 μL of enzyme (1 FDU) cleaves 1 μg of substrate DNA in 5-15 minutes in 1X FastDigest Buffer
> Hazardous:No
> Shipping Condition:Dry Ice
> Storage Condition:-20°C