missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fc gamma RIIA/CD32a Antibody (IV.3) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
267.00 € - 624.00 €
Spezifikation
| Antigen | Fc gamma RIIA/CD32a |
|---|---|
| Klon | IV.3 |
| Konzentration | 1 mg/mL |
| Anwendungen | Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Inhibition Assays |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18650294
|
Novus Biologicals
NBP3-20159-0.025mg |
0.025 mg |
267.00 €
0.02 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18657134
|
Novus Biologicals
NBP3-20159-0.2mg |
0.2 mg |
624.00 €
0.20 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Fc gamma RIIA/CD32a Monoclonal antibody specifically detects Fc gamma RIIA/CD32a in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Inhibition Activity, NeutralizationSpezifikation
| Fc gamma RIIA/CD32a | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD32 antigen, CD32A, CD32MGC23887, CDw32fc-gamma-RIIa, Fc fragment of IgG, low affinity IIa, receptor (CD32), Fc fragment of IgG, low affinity IIa, receptor for (CD32), FCG2, Fc-gamma RII-a, Fc-gamma-RIIa, FcGR, FCGR2, FCGR2A1, FcRII-a, IGFR2MGC30032, IgG Fc receptor II-a, Immunoglobulin G Fc receptor II, low affinity immunoglobulin gamma Fc region receptor II-a | |
| The original antibody was generated against the FcR of K562 cells. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| IV.3 | |
| Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Inhibition Assays | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS | |
| 2212 | |
| IgG κ | |
| Protein A purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts