Description
Fc gamma RIIB/CD32b Monoclonal antibody specifically detects Fc gamma RIIB/CD32b in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Fc gamma RIIB/CD32b |
| Anwendungen | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | 2E10 |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Zusammensetzung | In 1x PBS, pH 7.4 |
| Gen-Zugriffsnummer | AAH31992 |
| Gen-Alias | CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b |
| Wirtsspezies | Mouse |
| Show More |