Learn More
Fc gamma RIIB/CD32b Antibody (S07-4B5), Novus Biologicals™
Artikelnummer. 18687824 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
50 μg
Packungsgröße:
100 Mikroliter
50 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18687824 50 μg 50 Mikroliter
18623934 100 μg 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18687824 Lieferant Novus Biologicals Lieferanten-Nr. NBP31949650ul
Please to purchase this item. Need a web account? Register with us today!
Rabbit Monoclonal Antibody
Fc gamma RIIB/CD32b Monoclonal antibody specifically detects Fc gamma RIIB/CD32b in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Fc gamma RIIB/CD32b
Anwendungen Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon S07-4B5
Konzentration 0.3 mg/mL
Konjugat Unconjugated
Verdünnung Western Blot, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin
Zusammensetzung 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gen-Alias CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Wirtsspezies Rabbit
Immunogen A synthetic peptide of human Fc gamma RIIB/CD32b (Uniprot # P31994)
Reinigungsverfahren Affinity purified
Menge 50 μg
Regulatorischer Status RUO
Forschungsgebiet Immunology, Phospho Specific
Primär oder sekundär Primary
Gen-ID (Entrez) 2213
Zielspezies Human, Rat
Inhalt und Lagerung Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG