missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
FCAR/CD89 Monoclonal antibody specifically detects FCAR/CD89 in Human samples. It is validated for ELISA, ELISA
Spezifikation
Spezifikation
| Antigen | FCAR/CD89 |
| Anwendungen | ELISA |
| Klassifikation | Monoclonal |
| Klon | 002 |
| Konjugat | Alexa Fluor 700 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor |
| Wirtsspezies | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human FCAR/CD89 (NP_001991.1; Met1-Asn227). |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?