missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FDFT1 Monoclonal Antibody (24GB5645)
Mouse Monoclonal Antibody
Marke: Invitrogen™ MA557399
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FDFT1 Monoclonal Antibody for Western Blot
FDFT1 catalyzes the first step in the cholesterol biosynthetic pathway, the conversion of trans-farnesyldiphosphate to squalene. The loss of promoter activity and response to sterols for FDFT1 is localized to a 69-bp section positioned 131 bp 5-prime to the transcription start site. Sequence analysis of this region shows that it contains a sterol regulatory element-1 (SRE1) previously identified in other sterol regulated genes and 2 putative NF1 binding sites.
Spécification
| FDFT1 | |
| Monoclonal | |
| Unconjugated | |
| FDFT1 | |
| DGPT; EGK_18680; ERG9; farnesyl diphosphate farnesyl transferase 1; farnesyl diphosphate farnesyltransferase; farnesyl-diphosphate farnesyltransferase; farnesyl-diphosphate farnesyltransferase 1; FDFT1; FPP:FPP farnesyltransferase; presqualene-di-diphosphate synthase; SQS; Squalene synthase; squalene synthetase; SS | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2222 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5645 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P37268 | |
| FDFT1 | |
| Recombinant protein of human FDFT1 (aa 1-417). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu