Learn More
Invitrogen™ FDFT1 Monoclonal Antibody (24GB5645)
Mouse Monoclonal Antibody
Marke: Invitrogen™ MA557398
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
FDFT1 Monoclonal Antibody for Western Blot
FDFT1 catalyzes the first step in the cholesterol biosynthetic pathway, the conversion of trans-farnesyldiphosphate to squalene. The loss of promoter activity and response to sterols for FDFT1 is localized to a 69-bp section positioned 131 bp 5-prime to the transcription start site. Sequence analysis of this region shows that it contains a sterol regulatory element-1 (SRE1) previously identified in other sterol regulated genes and 2 putative NF1 binding sites.
Spezifikation
| FDFT1 | |
| Monoclonal | |
| Unconjugated | |
| FDFT1 | |
| DGPT; EGK_18680; ERG9; farnesyl diphosphate farnesyl transferase 1; farnesyl diphosphate farnesyltransferase; farnesyl-diphosphate farnesyltransferase; farnesyl-diphosphate farnesyltransferase 1; FDFT1; FPP:FPP farnesyltransferase; presqualene-di-diphosphate synthase; SQS; Squalene synthase; squalene synthetase; SS | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2222 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5645 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P37268 | |
| FDFT1 | |
| Recombinant protein of human FDFT1 (aa 1-417). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |