Learn More
Instech Laboratories™ Feeding tube, rat oral gavage, polyurethane, 13ga x 88mm, sterile
Beschreibung
These specialty tubes feature a larger inner diameter for easier delivery of viscous compounds and a straight tip that wipes clean to help in cases where aspiration of compounds would be hazardous to the animal. Availablein two sizes: 13ga for rats and 16ga for mice.
- Plastic feeding tubes, 13ga x 88mm
- For rat oral gavage
- Designed to minimise trauma to oesophagus
- Polyurethane tube with rounded tip
- Large internal diameter for viscous compounds
- Straight tube wipes clean to prevent aspiration of compounds
- Disposable
- Supplied as 10 sterile pouches of 5 tubes
Spezifikation
Spezifikation
| Produkttyp | Zulaufschlauch |