missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Feline IL-6 DuoSet ELISA
Click to view available options
Menge:
1 Kit (für 15 Platten)
Packungsgröße:
1 Set
Description
The Feline IL-6 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies feline IL-6.
Specifications
Specifications
| Menge | 1 Kit (für 15 Platten) |
| Spezies | Feline |
| Konjugat | Biotin |
| Assay-Bereich | 31.20 to 2000 pg/mL |
| Zur Verwendung mit (Anwendung) | ELISA |
| Gen-ID (Entrez) | 493687 |
| Synonym | B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A |
| Inhalt und Lagerung | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction