missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Felzartamab Recombinant Monoclonal Antibody

Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Felzartamab is a fully human recombinant immunoglobulin G1 (IgG1) monoclonal antibody (mAb) that targets CD38 on plasma cells. CD38 is a multifunctional enzyme that is widely expressed in both immune and non-hematopoietic cells. It plays a critical role in NAD+ metabolism by converting NAD+ to ADP-ribose and nicotinamide, which are key molecules in cellular signaling and energy metabolism. Structurally, CD38 is a transmembrane glycoprotein with ectoenzyme activity, meaning that its enzymatic functions occur on the outer surface of the cell membrane. CD38's activity is particularly significant in aging, as its expression increases in immune cells due to the senescence-associated secretory phenotype. This elevation in CD38 activity contributes to decreased NAD+ levels, which is associated with various age-related diseases. CD38 has also been identified as a potential therapeutic target for conditions such as renal immune diseases and age-related metabolic decline.
Spezifikation
Spezifikation
| Antigen | Felzartamab |
| Anwendungen | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Klassifikation | Recombinant Monoclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Wirtsspezies | Human |
| Reinigungsverfahren | Protein A |
| Menge | 100 μg |
| Regulatorischer Status | RUO |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?