missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Ferritin Monoclonal antibody specifically detects Ferritin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Ferritin |
| Anwendungen | Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | FTH/2082 |
| Konjugat | DyLight 350 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant human Ferritin protein fragment (around aa 58-180) (exact sequence is proprietary) (Uniprot: P02794) |
| Reinigungsverfahren | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?