Learn More
Ferritin mitochondrial Antibody (AMB8LK), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-43585
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Ferritin mitochondrial Monoclonal antibody specifically detects Ferritin mitochondrial in Human samples. It is validated for Immunoprecipitation
Spezifikation
| Ferritin mitochondrial | |
| Monoclonal | |
| 1 mg/mL | |
| Immunoprecipitation | |
| EC 1.16.3.1, ferritin H subunit, ferritin mitochondrial, ferritin, mitochondrial, mitochondrial ferritin, MTF | |
| The original antibody was generated by immunizing female Balb/c mice with ferritin extracted from human spleen cells. (Uniprot# Q8N4E7) | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 94033 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunoprecipitation | |
| AMB8LK | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |