Learn More
Fisherbrand™ Schellbach Burets with Straight Std. Taper Glass Stopcock
Provides accurate readings.
Marke:  Fisherbrand™ 03740C
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15102875
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
Provides accurate readings.
  • Accentuate meniscus for easy reading
  • Accuracy established within the limits of error allowed by NIST™