missing translation for 'onlineSavingsMsg'
Learn More

FITC Antibody, FITC , Novus Biologicals™

Artikelnummer. 18571112 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
1 mL
Unit Size:
1 Milliliter
Product Code. Menge unitSize
18571112 1 mL 1 Milliliter
1 options
This item is not returnable. View return policy

Product Code. 18571112

Brand: Novus Biologicals NB10064617F

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

FITC Polyclonal antibody specifically detects FITC in Nonspecies specific samples. It is validated for Flow Cytometry, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FITC
Anwendungen Flow Cytometry, ELISA
Klassifikation Polyclonal
Konjugat FITC
Zusammensetzung PBS
Gen-Alias Fluorescein Isothiocyanate
Wirtsspezies Sheep
Immunogen Pooled antisera from sheep hyperimmunized with fluorescein isothiocyanate (Isomer I)
Reinigungsverfahren Immunogen affinity purified
Menge 1 mL
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Non-species specific
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.