Learn More
Value Plastics Fitting Manifold with 200 Series Barbs, Polypropylene
Alle ansehen Cole-Parmer ProdukteBeschreibung
Constructed of ADCF polypropylene. Utilize 200 Series barbs in high pressure applications. Manufactured to strict quality standards under an ISO 9001:2000 quality control
Spezifikation
Spezifikation
| Zertifizierung/Konformität | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Verpackungsmenge | 25/Pk. |
| Auslassanschluss | Hose Barb Union Manifold |
| Sterilisationsmethode | 121°C |
| Form | Manifold |