missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FLAP Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA518283
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
ALOX5AP is a protein which, with 5-lipoxygenase, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.
Spezifikation
| FLAP | |
| Polyclonal | |
| Unconjugated | |
| ALOX5AP | |
| 5-lipoxygenase-activating protein FLAP; Alox5ap; arachidonate 5-lipoxygenase activating protein; arachidonate 5-lipoxygenase-activating protein; Flap; MK-886-binding protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 241 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P20292 | |
| ALOX5AP | |
| Synthetic peptide sequence (KTISTTISPLLLIP) corresponding to the C-terminus amino acids of ALOX5AP (aa 148-161). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur