missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flubendazole-d3 (methyl-d3), CDN
Click to view available options
Menge:
0.005 g
0.01 g
Conditionnement:
0.01 Gramm
Specifications
Specifications
| Chemischer Name oder Material | Flubendazole D3 |
| CAS | 1173021-08-3 |
| Summenformel | C16 2H3 H9 F N3 O3 |
| Menge | 0.005 g |
| Synonym | Trideuteriomethyl N-[5-(4-fluorobenzoyl)-1 H-benzimidazol-2-yl]carbamate, Flubendazole D3, Flubendazole D3 (methyl D3) |
| InChI-Formel | InChI=1 S/C16H12FN3O3/c1-23-16(22)20-15-18-12-7-4-10(8-13(12)19-15)14(21)9-2-5-11(17)6-3-9/h2-8 H,1H3,(H2,18,19,20,22)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])OC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccc(F)cc3 |
| IUPAC-Name | trideuteriomethyl N-[5-(4-fluorobenzoyl)-1 H-benzimidazol-2-yl]carbamate |
| Molekulargewicht (g/mol) | 316.3 |
| Formelmasse | 316.1051 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction