Learn More
Invitrogen™ FLYWCH1 Polyclonal Antibody
Beschreibung
Recommended positive controls: HeLa, HeLa nucleus. Predicted reactivity: Mouse (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Transcription cofactor. Negatively regulates transcription activation by catenin beta-1 CTNNB1, perhaps acting by competing with TCF4 for CTNNB1 binding. May play a role in DNA-damage response signaling. Binds specifically to DNA sequences at peri-centromeric chromatin loci.
Spezifikation
Spezifikation
| Antigen | FLYWCH1 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.61 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gen | FLYWCH1 |
| Gen-Zugriffsnummer | Q4VC44 |
| Gen-Alias | E030034P13Rik; FLYWCH1; FLYWCH-type zinc finger 1; FLYWCH-type zinc finger-containing protein 1; KIAA1552; mKIAA1552 |
| Gensymbole | FLYWCH1 |
| Mehr anzeigen |