Learn More
Folinic acid, MedChemExpress
Artikelnummer. 16467366
Encompass_Preferred
Change view
Click to view available options
Menge:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
16467366 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 16467366 Lieferant Medchem Express Lieferanten-Nr. HY17556
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
MedChemExpress Folinic acid (Leucovorin) is a biological folic acid and is generally administered along with Methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity.
Folinic acid (Leucovorin) is a biological folic acid and is generally administered along with Methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity.
Spezifikation
Chemischer Name oder Material Folinic acid
Physikalische Form Solid
CAS 58-05-9
Farbe Light Green
Menge 100 mg
Hinweise zur Reinheitsqualität Research
Summenformel C20H23N7O7
Zur Verwendung mit (Anwendung) Cancer-programmed cell death
Synonym leucovorin
Löslichkeitsinformationen DMSO : 250 mg/mL (528.05 mM; Need ultrasonic)
SMILES O=C(O)CC[C@@H](C(O)=O)NC(C1=CC=C(NCC2N(C=O)C3=C(N=C(N)NC3=O)NC2)C=C1)=O
Molekulargewicht (g/mol) 473.44
Formelmasse 473.44
Reinheit (%) 98.36%
Güte Research