missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Formyl-HIST1H2AG (Lys95) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5144294
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Formyl-HIST1H2AG (Lys95) Polyclonal Antibody for ICC/IF, ELISA
Histones are nuclear proteins that play an important role in maintaining the structure of the chromosome in eukaryotes. The nucleosome is made up of around 146bp of DNA, which enfolds around a histone octomer unit comprising of four major histones, H2A, H2B, H3 and H4. An important role of H2A is to ensure proper assembly or functioning of the centrometre. They also play an important role in gene regulation. The protein can be modified by acetylation and phosphorylation.
Spezifikation
| Formyl-HIST1H2AG (Lys95) | |
| Polyclonal | |
| Unconjugated | |
| H2AC11 | |
| H2A histone family, member P; H2A.1; H2A.1b; H2A/d; H2A/p; H2AC11; H2AC13; H2AC15; H2AC16; H2AC17; H2AFC; H2AFD; H2AFI; H2AFN; H2AFP; H2AG; H2ak; HIST1H2AG; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; histone 1, H2ag; histone cluster 1, H2ag; histone H2A type 1; histone H2A/e; histone H2A/p; Histone H2A/ptl; pH2A/f | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8329, 8969 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P0C0S8 | |
| H2AC11, HIST1H2AG | |
| Peptide sequence around site of Formyl-Lys (95) derived from Human Histone H2A type 1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur