missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Formyl-Histone H1.5 (Lys66) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5121408
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody tested in Indirect ELISA.
Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family.
Spezifikation
| Formyl-Histone H1.5 (Lys66) | |
| Polyclonal | |
| Unconjugated | |
| H1-5 | |
| H1; H1 histone family; H1 histone family, member 5; H1.5; H1B; H1F5; H1s-3; HIST1H1B; histone 1, H1b; histone cluster 1, H1b; histone H1.5; Histone H1a; histone H1b; Histone H1s-3; MGC126630; MGC126632 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3009 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 0.08 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P16401 | |
| H1-5, HIST1H1B | |
| Synthesized peptide derived from Human Histone H1.5 protein (63-75aa). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur