missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXA1 Monoclonal Antibody (654126)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561718
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver.
Spezifikation
| FOXA1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P55317 | |
| FOXA1 | |
| E. coli-derived recombinant human HNF-3 alpha /FoxA1, Met253-Ser472. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 654126 | |
| Unconjugated | |
| FOXA1 | |
| Ci-fkh; fkd7; fkh; fork head domain; forkhead box A1; forkhead box protein A1; forkhead-7; Foxa1; FoxA4a; hepatocyte nuclear factor 3 alpha (winged helix transcription factor); hepatocyte nuclear factor 3, alpha; hepatocyte nuclear factor 3-alpha; HNF3A; HNF-3A; HNF3alpha; HNF-3-alpha; HNF3-alpha; MGC33105; MGC52590; MGC75967; pintallavis; TCF3A; Tcf-3a; tFoxA1; Transcription factor 3A; xfkh1; xfkh2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3169 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur